(4-Methoxybenzyl)triphenylphosphonium chloride ≥95%

Supplier: Ambeed
Warning

A629110-1G A629110-25G A629110-5G
77602-590EA 10.25 USD
77602-590 77882-462 77882-463
(4-Methoxybenzyl)triphenylphosphonium chloride ≥95%
(4-Methoxybenzyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₆H₂₄ClOP
MDL Number: MFCD00031572
CAS Number: 3462-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR