Methyl 2-Boc-2-aza-spiro[3.3]heptane-6-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-tert-butyl 6-methyl 2-azaspiro[3.3]heptane-2,6-dicarboxylate

A629042-100MG A629042-250MG
77613-944EA 81.37 USD
77613-944 77613-946
Methyl 2-Boc-2-aza-spiro[3.3]heptane-6-carboxylate ≥98%
Methyl 2-Boc-2-aza-spiro[3.3]heptane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₄
MW: 255.31 g/mol
MDL Number: MFCD23106118
CAS Number: 1408074-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR