3-Chloro-2-nitrobenzyl alcohol ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A628957-250MG A628957-1G A628957-5G
77684-511EA 13 USD
77684-511 77893-265 77893-266
3-Chloro-2-nitrobenzyl alcohol ≥97%, Ambeed.Inc
3-Chloro-2-nitrobenzyl alcohol

Precaución: For research use only.

Formula: C₇H₆ClNO₃
MDL Number: MFCD04038814
CAS Number: 77158-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR