4-Bromo-1-(2-methoxyethyl)pyridin-2(1H)-one ≥95%

Supplier: Ambeed
Warning

A628666-100MG A628666-250MG
77617-654EA 124.63 USD
77617-654 77617-656
4-Bromo-1-(2-methoxyethyl)pyridin-2(1H)-one ≥95%
4-Bromo-1-(2-methoxyethyl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO₂
MDL Number: MFCD29038530
CAS Number: 1936135-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR