N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxamide ≥98%

Supplier: Ambeed
Warning

A628496-1G A628496-250MG A628496-100MG A628496-5G
77617-652EA 414.06 USD
77617-652 77749-283 77749-282 77749-284
N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxamide ≥98%
N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxamide

Precaución: for Research Use Only

Formula: C₁₂H₂₀BN₃O₃
MW: 265.12 g/mol
MDL Number: MFCD12405504
CAS Number: 942070-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR