(8S,9S,10R,13S,14S,17S)-17-((R)-2-Hydroxy-6-methylheptan-2-yl)-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol ≥98%

Supplier: Ambeed
Warning

A628488-100MG A628488-10MG A628488-50MG A628488-1MG A628488-250MG A628488-5MG
77617-646EA 546.93 USD
77617-646 77617-648 77617-650 77747-294 77747-295 77747-296
(8S,9S,10R,13S,14S,17S)-17-((R)-2-Hydroxy-6-methylheptan-2-yl)-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol ≥98%
(8S,9S,10R,13S,14S,17S)-17-((R)-2-Hydroxy-6-methylheptan-2-yl)-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol

Precaución: for Research Use Only

Formula: C₂₇H₄₆O₂
MDL Number: MFCD16661190
CAS Number: 516-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR