6-Bromobenzo[d]thiazole-4-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A627793-250MG A627793-1G
77749-279EA 433 USD
77749-279 77749-278
6-Bromobenzo[d]thiazole-4-carboxylic acid ≥95%
6-Bromobenzo[d]thiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂S
MW: 258.09 g/mol
MDL Number: MFCD31616746
CAS Number: 1784111-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR