2-(Hydroxymethyl)isonicotinic acid ≥98%

Supplier: Ambeed
Warning

A627241-100MG A627241-250MG
77886-509EA 98 USD
77886-509 77886-510
2-(Hydroxymethyl)isonicotinic acid ≥98%
2-(Hydroxymethyl)isonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MDL Number: MFCD11977439
CAS Number: 915140-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR