Nitrazine yellow BS

Supplier: Ambeed
Danger

Synonyms: Sodium 3-((2,4-dinitrophenyl)diazenyl)-4-hydroxynaphthalene-2,7-disulphonate

A626464-1G A626464-25G A626464-5G
77669-698EA 7 USD
77669-698 77880-333 77880-334
Nitrazine yellow BS
Nitrazine yellow

Precaución: for Research Use Only

Formula: C₁₆H₈N₄Na₂O₁₁S₂
MW: 542.36 g/mol
MDL Number: MFCD00003938
CAS Number: 5423-07-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR