2-Acetamido-3-mercaptopropanoic acid ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-Ac-DL-Cys-OH

A626373-10G A626373-1G A626373-25G A626373-5G
77684-508EA 83 USD
77684-508 77891-238 77891-239 77891-240
2-Acetamido-3-mercaptopropanoic acid ≥97%, Ambeed.Inc
2-Acetamido-3-mercaptopropanoic acid

Precaución: For research use only.

Formula: C₅H₉NO₃S
MDL Number: MFCD00151956
CAS Number: 7218-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR