1-(Oxazol-4-yl)ethanone ≥96%

Supplier: Ambeed
Warning

A626009-100MG A626009-250MG
77749-275EA 54 USD
77749-275 77749-276
1-(Oxazol-4-yl)ethanone ≥96%
1-(Oxazol-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₅H₅NO₂
MDL Number: MFCD18250706
CAS Number: 859046-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR