7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid ≥95%

Supplier: Ambeed
Warning

A625982-5G A625982-100G A625982-1G
77602-580EA 76.88 USD
77602-580 77669-694 77669-696
7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid ≥95%
7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₉ClFNO₃
MDL Number: MFCD00792459
CAS Number: 68077-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR