2-Ethyl-1,1-dimethyl-1H-benzo[e]indole ≥95%

Supplier: Ambeed
Warning

A606900-250MG A606900-1G
77749-247EA 44 USD
77749-247 77749-246
2-Ethyl-1,1-dimethyl-1H-benzo[e]indole ≥95%
2-Ethyl-1,1-dimethyl-1H-benzo[e]indole

Precaución: for Research Use Only

Formula: C₁₆H₁₇N
CAS Number: 1092836-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR