tert-Butyl 2,6-diazaspiro[3.5]nonane-6-carboxylate ≥97%

Supplier: Ambeed
Warning

A606451-250MG A606451-1G A606451-5G A606451-100MG
77669-624EA 160 USD
77669-624 77880-299 77880-300 77880-298
tert-Butyl 2,6-diazaspiro[3.5]nonane-6-carboxylate ≥97%
tert-Butyl 2,6-diazaspiro[3.5]nonane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD08234981
CAS Number: 885272-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR