3-Chloropropanesulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A606018-25G A606018-5G
77572-962EA 45 USD
77572-962 77572-964
3-Chloropropanesulfonyl chloride ≥98%
3-Chloropropanesulfonyl chloride

Precaución: for Research Use Only

Formula: C₃H₆Cl₂O₂S
MDL Number: MFCD00007463
CAS Number: 1633-82-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR