(S)-tert-Butyl 2-aminopropanoate ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Ala-OtBu
A605887-1G
A605887-100MG
77594-848EA
62.53
USD
77594-848
77673-636
(S)-tert-Butyl 2-aminopropanoate ≥95%
(S)-tert-Butyl 2-aminopropanoate
Precaución: for Research Use Only
Formula:
C₇H₁₅NO₂ MW: 145.19 g/mol |
MDL Number:
MFCD00270591 CAS Number: 21691-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...