PLX-7904 98+%

Supplier: Ambeed
Warning

Synonyms: PLX7904, N'-[3-[[5-(2-Cyclopropyl-5-pyrimidinyl)-1H-pyrrolo[2,3-b]pyridin-3-yl]carbonyl]-2,4-difluorophenyl]-N-ethyl-N-methylsulphamide, 5-(2-Cyclopropylpyrimidin-5-yl)-3-[3-[[ethyl(methyl)sulphamoyl]amino]-2,6-difluorobenzoyl]-1H-pyrrolo[2,3-b]pyridine

A605818-2MG A605818-10MG A605818-5MG
77747-249EA 47 USD
77747-249 77747-248 77747-250
PLX-7904 98+%
PLX-7904

Precaución: for Research Use Only

Formula: C₂₄H₂₂F₂N₆O₃S
MDL Number: MFCD29472265
CAS Number: 1393465-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR