4-Nitroquinoline-N-oxide ≥98%

Supplier: Ambeed
Danger

A605439-1G A605439-250MG A605439-5G
77617-534EA 36.05 USD
77617-534 77617-536 77617-538
4-Nitroquinoline-N-oxide ≥98%
4-Nitroquinoline-N-oxide

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MW: 190.1555 g/mol
MDL Number: MFCD00006738
CAS Number: 56-57-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR