Methyl 5-(chlorosulfonyl)pentanoate ≥98%

Supplier: Ambeed
Danger

Synonyms: Methyl 5-(Chlorosulphonyl)pentanoate

A604094-250MG A604094-100MG A604094-1G A604094-5G
77613-874EA 127.72 USD
77613-874 77673-624 77673-626 77884-452
Methyl 5-(chlorosulfonyl)pentanoate ≥98%
Methyl 5-(chlorosulfonyl)pentanoate

Precaución: for Research Use Only

Formula: C₆H₁₁ClO₄S
MW: 214.667 g/mol
MDL Number: MFCD22574974
CAS Number: 1408058-14-9
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR