2,4-Di-tert-butoxy-5-pyrimidineboronic acid ≥98%

Supplier: Ambeed
Warning

A603758-250MG A603758-1G
77686-528EA 34 USD
77686-528 77893-219
2,4-Di-tert-butoxy-5-pyrimidineboronic acid ≥98%
2,4-Di-tert-butoxy-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁BN₂O₄
MDL Number: MFCD01318980
CAS Number: 109299-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR