1-Methyl-3-(3-methyl-4-(4-((trifluoromethyl)sulfonyl)phenoxy)phenyl)-1,3,5-triazinane-2,4,6-trione ≥98%

Supplier: Ambeed
Warning

A603396-10MG A603396-5MG A603396-100MG A603396-1G A603396-250MG A603396-50MG
77669-610EA 39.06 USD
77669-610 77669-612 77880-283 77880-284 77880-285 77880-286
1-Methyl-3-(3-methyl-4-(4-((trifluoromethyl)sulfonyl)phenoxy)phenyl)-1,3,5-triazinane-2,4,6-trione ≥98%
1-Methyl-3-(3-methyl-4-(4-((trifluoromethyl)sulfonyl)phenoxy)phenyl)-1,3,5-triazinane-2,4,6-trione

Precaución: for Research Use Only

Formula: C₁₈H₁₄F₃N₃O₆S
MDL Number: MFCD09263475
CAS Number: 69004-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR