Tyrphostin B42 ≥98%

Supplier: Ambeed
Danger

A603326-10MG A603326-1MG A603326-5MG
77602-516EA 48.18 USD
77602-516 77602-518 77602-520
Tyrphostin B42 ≥98%
Tyrphostin B42

Precaución: for Research Use Only

Formula: C₁₇H₁₄N₂O₃
MDL Number: MFCD00236452
CAS Number: 134036-52-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR