1-Iodo-4,4,4-trifluorobutane ≥98%

Supplier: Ambeed
Danger

Synonyms: 4-Iodo-1,1,1-trifluorobutane, 1,1,1-Trifluoro-4-iodobutane

A603313-250MG A603313-10G
77749-240EA 8 USD
77749-240 77749-239
1-Iodo-4,4,4-trifluorobutane ≥98%
1-Iodo-4,4,4-trifluorobutane

Precaución: for Research Use Only

Formula: C₄H₆F₃I
MW: 237.99 g/mol
MDL Number: MFCD00041543
CAS Number: 461-17-6
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR