5-Chlorobenzo[b]thiophene-3-carboxaldehyde ≥95%

Supplier: Ambeed
Warning

A603189-1G A603189-250MG
77878-400EA 183 USD
77878-400 77878-401
5-Chlorobenzo[b]thiophene-3-carboxaldehyde ≥95%
5-Chlorobenzo[b]thiophene-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₅ClOS
MDL Number: MFCD02677690
CAS Number: 16296-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR