5-Fluoro-2-(6-fluoro-2-methyl-1H-benzo[d]imidazol-1-yl)-N4-(4-(trifluoromethyl)phenyl)pyrimidine-4,6-diamine 98+%

Supplier: Ambeed
Warning

Synonyms: Unesbulin

A602809-1MG A602809-5MG
77617-522EA 11.33 USD
77617-522 77617-524
5-Fluoro-2-(6-fluoro-2-methyl-1H-benzo[d]imidazol-1-yl)-N4-(4-(trifluoromethyl)phenyl)pyrimidine-4,6-diamine 98+%
5-Fluoro-2-(6-fluoro-2-methyl-1H-benzo[d]imidazol-1-yl)-N4-(4-(trifluoromethyl)phenyl)pyrimidine-4,6-diamine

Precaución: for Research Use Only

Formula: C₁₉H₁₃F₅N₆
MW: 420.34 g/mol
MDL Number: MFCD30489439
CAS Number: 1610964-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR