Dimethyl (2-oxo-3-phenoxypropyl)phosphonate ≥98%

Supplier: Ambeed
Warning

A602463-250MG A602463-5G
77880-280EA 8 USD
77880-280 77880-281
Dimethyl (2-oxo-3-phenoxypropyl)phosphonate ≥98%
Dimethyl (2-oxo-3-phenoxypropyl)phosphonate

Precaución: for Research Use Only

Formula: C₁₁H₁₅O₅P
MDL Number: MFCD00010225
CAS Number: 40665-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR