Fmoc-4-(trifluoromethyl)-D-phenylalanine ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(4-CF3)-OH, Fmoc-D-Phe(4-CF₃)-OH

A602072-1G A602072-250MG
77882-412EA 90 USD
77882-412 77882-413
Fmoc-4-(trifluoromethyl)-D-phenylalanine ≥98%
Fmoc-4-(trifluoromethyl)-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₅H₂₀F₃NO₄
MW: 455.44 g/mol
MDL Number: MFCD00797582
CAS Number: 238742-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR