2,5-Difluoro-3-nitrobenzoic acid ≥98%

Supplier: Ambeed
Warning

A601510-100MG A601510-250MG
77891-190EA 40 USD
77891-190 77891-191
2,5-Difluoro-3-nitrobenzoic acid ≥98%
2,5-Difluoro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₂NO₄
MDL Number: MFCD11848537
CAS Number: 741721-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR