tert-Butyl 1,6-diazaspiro[3.4]octane-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A601262-1G A601262-250MG
77572-940EA 823 USD
77572-940 77572-942
tert-Butyl 1,6-diazaspiro[3.4]octane-1-carboxylate ≥98%
tert-Butyl 1,6-diazaspiro[3.4]octane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD12406507
CAS Number: 1148044-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR