3,5,7-Trihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one 98+%

Supplier: Ambeed
Warning

Synonyms: Tamarixetin

A601013-1MG A601013-10MG A601013-5MG
77602-510EA 76.88 USD
77602-510 77669-602 77669-604
3,5,7-Trihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one 98+%
3,5,7-Trihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₇
MW: 316.26 g/mol
MDL Number: MFCD00017308
CAS Number: 603-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR