4,7-Dichloropyrido[3,2-d]pyrimidine ≥97%

Supplier: Ambeed
Warning

A600625-100MG A600625-1G
77686-522EA 81 USD
77686-522 77686-523
4,7-Dichloropyrido[3,2-d]pyrimidine ≥97%
4,7-Dichloropyrido[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
MW: 200.02 g/mol
MDL Number: MFCD18260280
CAS Number: 917757-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR