Disodium sebacate ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium decanedioate, Decanedioic Acid Disodium Salt, Disodium Decanedioate, Disodium Sebacate, Sebacic Acid Disodium Salt

A600526-1000G A600526-100G A600526-25G A600526-500G
77572-936EA 57 USD
77572-936 77602-502 77602-504 77602-506
Disodium sebacate ≥98%
Disodium sebacate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MDL Number: MFCD00070501
CAS Number: 17265-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR