5-Amino-1-naphthol ≥97%

Supplier: Ambeed
Warning

A600469-10G A600469-5G
77884-446EA 35 USD
77884-446 77884-447
5-Amino-1-naphthol ≥97%
5-Amino-1-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD00041826
CAS Number: 83-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR