3-Bromo-5-fluoro-2-iodotoluene ≥95%

Supplier: Ambeed
Warning

A600149-1G A600149-25G A600149-5G
77900-995EA 39 USD
77900-995 77901-460 77901-543
3-Bromo-5-fluoro-2-iodotoluene ≥95%
3-Bromo-5-fluoro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MDL Number: MFCD09800772
CAS Number: 1000576-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR