D-(+)-Glucose-6-phosphate sodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: ROBISON ESTER MONOSODIUM SALT, Sodium (2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl hydrogenphosphate, G-6-P Na, BETA-D-GLUCOSE 6-PHOSPHATE MONOSODIUM SALT, D-Glucose 6-phosphate sodium

A600123-10G A600123-250MG A600123-5G
77669-598EA 617 USD
77669-598 77669-600 77872-988
D-(+)-Glucose-6-phosphate sodium salt ≥98%
D-(+)-Glucose-6-phosphate sodium salt

Precaución: for Research Use Only

Formula: C₆H₁₂NaO₉P
MW: 282.12 g/mol
MDL Number: MFCD00006611
CAS Number: 54010-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR