RS-127445 hydrochloride ≥98%

Supplier: Ambeed
Danger

Synonyms: RS-127445 HCl, 4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine hydrochloride

A599861-5MG A599861-10MG
77613-868EA 11.33 USD
77613-868 77673-612
RS-127445 hydrochloride ≥98%
RS-127445 hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₆FN₃·HCl
MW: 317.79 g/mol
MDL Number: MFCD11112196
CAS Number: 199864-86-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR