3-(3-Fluorophenyl)-1H-pyrazol-5-amine ≥98%

Supplier: Ambeed
Warning

A599647-1G A599647-250MG
77891-187EA 62 USD
77891-187 77891-188
3-(3-Fluorophenyl)-1H-pyrazol-5-amine ≥98%
3-(3-Fluorophenyl)-1H-pyrazol-5-amine

Precaución: for Research Use Only

Formula: C₉H₈FN₃
MDL Number: MFCD06738760
CAS Number: 766519-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR