2-(6-Fluoropyridin-2-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

A599571-1G A599571-250MG
77893-210EA 262 USD
77893-210 77893-211
2-(6-Fluoropyridin-2-yl)acetic acid ≥98%
2-(6-Fluoropyridin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD09925118
CAS Number: 1000517-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR