Potassium(4-ethoxyphenyl)trifluoroborate ≥97%

Supplier: Ambeed
Warning

Synonyms: Potassium (4-ethoxyphenyl)trifluoroborate, (4-Ethoxyphenyl)trifluoroboric acid potassium salt

A599444-1G A599444-25G A599444-5G
77880-274EA 23 USD
77880-274 77880-275 77880-276
Potassium(4-ethoxyphenyl)trifluoroborate ≥97%
Potassium(4-ethoxyphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₈H₉BF₃KO
MW: 228.06
MDL Number: MFCD04115753
CAS Number: 850623-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR