1-((2R,3R)-3-(4-(4-Cyanophenyl)thiazol-2-yl)-2-(2,5-difluorophenyl)-2-hydroxybutyl)-4-(1-((methyl(3-(((methylglycyl)oxy)methyl)pyridin-2-yl)carbamoyl)oxy)ethyl)-1H-1,2,4-triazol-4-ium hydrogen sulfate ≥98%

Supplier: Ambeed
Warning

Synonyms: Isavuconazonium sulphate

A599121-100MG A599121-250MG
77617-510EA 127.72 USD
77617-510 77617-512
1-((2R,3R)-3-(4-(4-Cyanophenyl)thiazol-2-yl)-2-(2,5-difluorophenyl)-2-hydroxybutyl)-4-(1-((methyl(3-(((methylglycyl)oxy)methyl)pyridin-2-yl)carbamoyl)oxy)ethyl)-1H-1,2,4-triazol-4-ium hydrogen sulfate ≥98%
1-((2R,3R)-3-(4-(4-Cyanophenyl)thiazol-2-yl)-2-(2,5-difluorophenyl)-2-hydroxybutyl)-4-(1-((methyl(3-(((methylglycyl)oxy)methyl)pyridin-2-yl)carbamoyl)oxy)ethyl)-1H-1,2,4-triazol-4-ium hydrogen sulfate

Precaución: for Research Use Only

Formula: C₃₅H₃₆F₂N₈O₉S₂
MW: 814.84 g/mol
MDL Number: MFCD27987905
CAS Number: 946075-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR