5-Methyl-1H-indole-2-carbaldehyde ≥98%

Supplier: Ambeed
Warning

A589053-100MG A589053-1G A589053-250MG
77882-396EA 29 USD
77882-396 77882-397 77882-398
5-Methyl-1H-indole-2-carbaldehyde ≥98%
5-Methyl-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD06801043
CAS Number: 1463-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR