(6R,7R)-7-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate ≥97%

Supplier: Ambeed
Danger

Synonyms: Cefadroxil hydrate

A588971-1MG A588971-10MG A588971-50MG
77747-222EA 9 USD
77747-222 77747-221 77747-223
(6R,7R)-7-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate ≥97%
(6R,7R)-7-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₃O₆S
MDL Number: MFCD01682047
CAS Number: 66592-87-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR