Siponimod (BAF-312) ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-Azetidinecarboxylic acid, Siponimod, 1-{4-[1-((E)-4-cyclohexyl-3-trifluoromethyl-benzyloxyimino)-ethyl]-2-ethyl-benzyl}-azetidine-3-carboxylic acid, 1-[[4-[(E)-N-[[4-cyclohexyl-3-(trifluoromethyl)phenyl]methoxy]-C-methylcarbonimidoyl]-2-ethylphenyl]methyl]azetidine-3-carboxylic acid, (E)-1-(4-(1-(((4-Cyclohexyl-3-(trifluoromethyl)benzyl)oxy)imino)ethyl)-2-ethylbenzyl)azetidine-3-carboxylic acid, 1-((4-((1E)-1-(((4-cyclohexyl-3-(trifluoromethyl)phenyl)methoxy)imino)ethyl)-2-ethylphenyl)methyl)

A588237-100MG A588237-50MG A588237-5MG
77613-826EA 361.53 USD
77613-826 77686-494 77686-495
Siponimod (BAF-312) ≥98%
Siponimod (BAF-312)

Precaución: for Research Use Only

Formula: C₂₉H₃₅F₃N₂O₃
MW: 516.6 g/mol
MDL Number: MFCD26142651
CAS Number: 1230487-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR