PD318088 ≥98%

Supplier: Ambeed
Warning

Synonyms: PD-318088, 5-Bromo-N-(2,3-dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamide, pd 318088, PD318088, pd318088, PD 318088, 5-bromo-N-(2,3-dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodoanilino)benzamide

A588037-1MG A588037-10MG A588037-5MG
77617-470EA 32.96 USD
77617-470 77686-491 77686-492
PD318088 ≥98%
PD318088

Precaución: for Research Use Only

Formula: C₁₆H₁₃BrF₃IN₂O₄
MW: 561.09 g/mol
MDL Number: MFCD18384973
CAS Number: 391210-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR