5-(Hydroxymethyl)pyridine-3-boronic Acid ≥98%

Supplier: Ambeed
Warning

A588000-100MG A588000-1G A588000-250MG A588000-5G
77627-222EA 32.96 USD
77627-222 77878-388 77878-389 77878-390
5-(Hydroxymethyl)pyridine-3-boronic Acid ≥98%
5-(Hydroxymethyl)pyridine-3-boronic Acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₃
MW: 152.94 g/mol
MDL Number: MFCD09952042
CAS Number: 908369-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR