N-(1-((2R,6S)-6-(Hydroxymethyl)-4-tritylmorpholin-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)benzamide ≥98% (contains ~10% solvents)

Supplier: Ambeed
Warning

Synonyms: N4-Benzoyl-7’-OH-N-trityl morpholino cytosine

A587460-100MG A587460-250MG
77747-217EA 23 USD
77747-217 77747-218
N-(1-((2R,6S)-6-(Hydroxymethyl)-4-tritylmorpholin-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)benzamide ≥98% (contains ~10% solvents)
N-(1-((2R,6S)-6-(Hydroxymethyl)-4-tritylmorpholin-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)benzamide

Precaución: for Research Use Only

Formula: C₃₅H₃₂N₄O₄
MW: 572.65 g/mol
MDL Number: MFCD22666302
CAS Number: 125515-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR