Methyl 4-(2-oxoethyl)bicyclo[2.2.1]heptane-1-carboxylate ≥97%

Supplier: Ambeed
Warning

A587453-100MG A587453-1G A587453-250MG
77783-480EA 143 USD
77783-480 77783-481 77783-482
Methyl 4-(2-oxoethyl)bicyclo[2.2.1]heptane-1-carboxylate ≥97%
Methyl 4-(2-oxoethyl)bicyclo[2.2.1]heptane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
MDL Number: MFCD31705162
CAS Number: 2231673-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR