6-Chloro-9-phenyl-9H-purine ≥95%

Supplier: Ambeed
Warning

A587196-100MG A587196-1G A587196-250MG
77891-164EA 27 USD
77891-164 77891-165 77891-166
6-Chloro-9-phenyl-9H-purine ≥95%
6-Chloro-9-phenyl-9H-purine

Precaución: for Research Use Only

Formula: C₁₁H₇ClN₄
MDL Number: MFCD16619574
CAS Number: 5470-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR