Caspofungin acetate ≥98%

Supplier: Ambeed
Warning

A587048-100MG A587048-1MG A587048-250MG A587048-5MG A587048-10MG A587048-1G
77602-454EA 104.55 USD
77602-454 77613-818 77686-487 77686-488 77900-914 77900-887
Caspofungin acetate ≥98%
Caspofungin acetate

Precaución: for Research Use Only

Formula: C₅₂H₈₈N₁₀O₁₅·₂C₂HO₂
MDL Number: MFCD08141839
CAS Number: 179463-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR