(4-fluoro-3-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A624068-100MG A624068-1G A624068-250MG
77893-251EA 143 USD
77893-251 77893-252 77893-253
(4-fluoro-3-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid 98+%
(4-fluoro-3-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BFNO₄S
MDL Number: MFCD28805612
CAS Number: 1704121-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR